Assay Details
| Gene Symbol: D3Ertd254e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101478.1 | NP_001094948.1 | 1 - 2 | 113 | 79 | |
| GenBank mRNA | AK153849.1 | - | 1 - 2 | 62 | 79 | |
| BB619095.1 | - | 1 - 2 | 115 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001101478.1 transcripts.
Set Membership: |
|

D3Ertd254e
Genomic Map