Assay Details
| Gene Symbol: D3Ertd254e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101478.1 | NP_001094948.1 | 2 - 3 | 243 | 117 | |
| GenBank mRNA | AK153849.1 | - | 2 - 3 | 192 | 117 | |
| BB619095.1 | - | 2 - 3 | 245 | 117 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D3Ertd254e
Genomic Map