Assay Details
| Gene Symbol: D3Ertd254e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101478.1 | NP_001094948.1 | 3 - 4 | 346 | 63 | |
| GenBank mRNA | AK153849.1 | - | 3 - 4 | 295 | 63 | |
| BB619095.1 | - | 3 - 4 | 348 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P511386 | AK033830.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P511386 | NM_001101478.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P511386 | AK033830.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P511386 | NM_001101478.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430690 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664768 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 161425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703012 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618628 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646421 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 54658 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42485 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117916 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P511386 | AK033830.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P511386 | NM_001101478.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P511386 | AK033830.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P511386 | NM_001101478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P328597 | AK153849.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P328597 | BB619095.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P328597 | NM_001101478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511386 | AK033830.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511386 | NM_001101478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511386 | AK033830.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511386 | NM_001101478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P487281 | AK153849.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P487281 | NM_001101478.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22056775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02942506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02713990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03146610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03146609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02550580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02766631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03088910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22056766 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1455757_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437557_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437557_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455757_at | - | - |
| Mouse Genome 430 2.0 Array | 1455757_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437557_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 241944_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 241944_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300001764.mm.1 | NM_001101478.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001764.mm.1 | NM_001101478.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001764.mm.1 | TC0300000266.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300001763.mm.1 | NM_001101478.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001763.mm.1 | NM_001101478.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001763.mm.1 | TC0300000266.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300001759.mm.1 | NM_001101478.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001759.mm.1 | NM_001101478.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001759.mm.1 | TC0300000266.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300001761.mm.1 | NM_001101478.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001761.mm.1 | NM_001101478.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001761.mm.1 | TC0300000266.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300001760.mm.1 | NM_001101478.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001760.mm.1 | NM_001101478.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001760.mm.1 | TC0300000266.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300001762.mm.1 | NM_001101478.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001762.mm.1 | NM_001101478.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300001762.mm.1 | TC0300000266.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455757_at | - | - |
| Mouse Expression Set 430 | 1437557_PM_at | - | - |
| Mouse Expression Set 430 | 1455757_PM_at | - | - |
| Mouse Expression Set 430 | 1437557_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437557_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455757_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455757_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437557_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 506713 | - | - |
| ABI Mouse Genome Survey Microarray | 531320 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101780324 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2475022 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2528305 | - | - |

D3Ertd254e
Genomic Map