Assay Details
| Gene Symbol: Mgst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174995.2 | NP_778160.2 | 2 - 3 | 213 | 88 | |
| GenBank mRNA | AA919832.1 | - | 2 - 3 | 213 | 88 | |
| BC132234.1 | - | 2 - 3 | 192 | 40130511 | 88 | |
| BC132260.1 | - | 2 - 3 | 192 | 40130537 | 88 | |
| BC145529.1 | - | 2 - 3 | 192 | 88 | ||
| BX639527.1 | - | 3 - 4 | 88 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Mgst2
Genomic Map