Assay Details
| Gene Symbol: Skil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039090.2 | NP_001034179.1 | 4 - 5 | 2005 | 95 | |
| NM_001271772.1 | NP_001258701.1 | 6 - 7 | 1686 | 95 | ||
| GenBank mRNA | AK154471.1 | - | 4 - 5 | 1952 | 95 | |
| AK171402.1 | - | 4 - 5 | 1954 | 95 | ||
| BC018600.1 | - | 6 - 7 | 1618 | 95 | ||
| BC049934.1 | - | 3 - 4 | 1431 | 5387475 | 95 | |
| U10530.1 | - | 3 - 4 | 1776 | 95 | ||
| U14655.1 | - | 3 - 4 | 1338 | 95 | ||
More Information
Set Membership: |
|

Skil
Genomic Map