Assay Details
Gene Symbol: Sec62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027016.2 | NP_081292.1 | 2 - 3 | 617 | 92 | |
GenBank mRNA | AK013922.1 | - | 2 - 3 | 432 | 92 | |
AK088093.1 | - | 2 - 3 | 166 | 92 | ||
AK090078.1 | - | 2 - 3 | 167 | 92 | ||
BC017162.1 | - | 2 - 3 | 618 | 92 | ||
BC048532.1 | - | 2 - 3 | 167 | 92 | ||
BC053504.1 | - | 2 - 3 | 154 | 92 | ||
BC067202.1 | - | 2 - 3 | 152 | 6332701 | 92 | |
BC071243.1 | - | 2 - 3 | 167 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
