Assay Details
| Gene Symbol: Egfem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029412.1 | NP_083688.1 | 8 - 9 | 1127 | 90 | |
| GenBank mRNA | AK032017.1 | - | 8 - 9 | 971 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Egfem1
Genomic Map