Assay Details
| Gene Symbol: Zfp639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161818.1 | NP_001155290.1 | 3 - 4 | 295 | 129 | |
| NM_144519.4 | NP_653102.1 | 3 - 4 | 396 | 129 | ||
| GenBank mRNA | AK049934.1 | - | 3 - 4 | 275 | 129 | |
| AK076145.1 | - | 3 - 4 | 397 | 129 | ||
| AK076837.1 | - | 3 - 4 | 272 | 129 | ||
| AK078000.1 | - | 3 - 4 | 296 | 129 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049934.1, AK076145.1, AK076837.1, AK078000.1, NM_001161818.1, NM_144519.4 transcripts.
Set Membership: |
|

Zfp639
Genomic Map