Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300003500.mm.1 |
NM_001033300.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003500.mm.1 |
NM_001033300.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003500.mm.1 |
TC0300000507.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300003502.mm.1 |
NM_001033300.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003502.mm.1 |
NM_001033300.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003502.mm.1 |
TC0300000507.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300003503.mm.1 |
NM_001033300.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003503.mm.1 |
NM_001033300.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003503.mm.1 |
TC0300000507.mm.1 |
MTA TC |