Assay Details
| Gene Symbol: Eif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005509.2 | NP_001005509.1 | 8 - 9 | 932 | 98 | |
| GenBank mRNA | AK044865.1 | - | 2 - 3 | 979 | 98 | |
| AK050971.1 | - | 8 - 9 | 649 | 98 | ||
| AK078519.1 | - | 8 - 9 | 711 | 98 | ||
| AK135515.1 | - | 2 - 3 | 1930 | 98 | ||
| AK145275.1 | - | 8 - 9 | 888 | 98 | ||
| BC030447.1 | - | 7 - 8 | 601 | 5400637 | 98 | |
| BC082557.1 | - | 8 - 9 | 737 | 30606936 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif2a
Genomic Map