Assay Details
| Gene Symbol: Uchl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019562.2 | NP_062508.2 | 8 - 9 | 855 | 173 | |
| GenBank mRNA | AF148447.1 | - | 8 - 9 | 798 | 173 | |
| AK011117.1 | - | 8 - 9 | 822 | 173 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uchl5
Genomic Map