Assay Details
| Gene Symbol: Ivns1abp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039512.1 | NP_001034601.1 | 8 - 9 | 1266 | 63 | |
| NM_054102.2 | NP_473443.2 | 9 - 10 | 1392 | 63 | ||
| GenBank mRNA | AB055737.1 | - | 9 - 10 | 1388 | 63 | |
| AK129229.1 | - | 9 - 10 | 1330 | 63 | ||
| AK144619.1 | - | 9 - 10 | 1372 | 63 | ||
| AK144835.1 | - | 8 - 9 | 1247 | 63 | ||
| AK145071.1 | - | 9 - 10 | 1383 | 63 | ||
| AK145845.1 | - | 9 - 10 | 1342 | 63 | ||
| AK146001.1 | - | 9 - 10 | 1373 | 63 | ||
| AK146494.1 | - | 9 - 10 | 1345 | 63 | ||
| AK146645.1 | - | 9 - 10 | 1343 | 63 | ||
| AK159254.1 | - | 9 - 10 | 1374 | 63 | ||
| AK197779.1 | - | 3 - 4 | 325 | 63 | ||
| BC004092.1 | - | 8 - 9 | 1092 | 3593827 | 63 | |
| DQ914522.1 | - | 9 - 10 | 1224 | 63 | ||
| DQ914523.1 | - | 9 - 10 | 2275 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ivns1abp
Genomic Map