Assay Details
| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008567.1 | NP_032593.1 | 5 - 6 | 875 | 90 | |
| GenBank mRNA | AK088493.1 | - | 5 - 6 | 874 | 90 | |
| AK145520.1 | - | 5 - 6 | 866 | 90 | ||
| AK146272.1 | - | 5 - 6 | 872 | 90 | ||
| AK146770.1 | - | 5 - 6 | 867 | 90 | ||
| AK150726.1 | - | 5 - 6 | 823 | 90 | ||
| AK185166.1 | - | 1 - 2 | 121 | 90 | ||
| BC050886.1 | - | 5 - 6 | 883 | 5703922 | 90 | |
| BC057584.1 | - | 5 - 6 | 853 | 6401827 | 90 | |
| D86726.1 | - | 5 - 6 | 875 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm6
Genomic Map