Assay Details
| Gene Symbol: Cadm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036352.1 | - | 1 - 2 | 236 | 140 | |
| BC079837.1 | - | 1 - 2 | 220 | 140 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cadm3| Gene Symbol: Cadm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036352.1 | - | 1 - 2 | 236 | 140 | |
| BC079837.1 | - | 1 - 2 | 220 | 140 | ||
Set Membership: |
|