Assay Details
| Gene Symbol: Cadm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036352.1 | - | 2 - 3 | 341 | 88 | |
| BC079837.1 | - | 2 - 3 | 325 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cadm3| Gene Symbol: Cadm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036352.1 | - | 2 - 3 | 341 | 88 | |
| BC079837.1 | - | 2 - 3 | 325 | 88 | ||
Set Membership: |
|