Assay Details
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023041.3 | NP_075528.3 | 2 - 3 | 205 | 103 | |
| GenBank mRNA | AK012785.1 | - | 2 - 3 | 188 | 103 | |
| AK029368.1 | - | 2 - 3 | 206 | 103 | ||
| AK075576.1 | - | 2 - 3 | 192 | 103 | ||
| BC012517.1 | - | 2 - 3 | 140 | 103 | ||
| BC019767.1 | - | 2 - 3 | 188 | 4502819 | 103 | |
| CT010305.1 | - | 2 - 3 | 181 | 103 | ||
| Y09046.1 | - | 2 - 3 | 186 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pex19
Genomic Map