Assay Details
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159525.1 | NP_001152997.1 | 2 - 3 | 181 | 87 | |
| NM_023041.3 | NP_075528.3 | 4 - 5 | 457 | 87 | ||
| GenBank mRNA | AK012785.1 | - | 4 - 5 | 440 | 87 | |
| AK028449.1 | - | 2 - 3 | 175 | 87 | ||
| AK029368.1 | - | 4 - 5 | 458 | 87 | ||
| AK075576.1 | - | 4 - 5 | 444 | 87 | ||
| AK170175.1 | - | 4 - 5 | 446 | 87 | ||
| BC012517.1 | - | 4 - 5 | 392 | 87 | ||
| BC019767.1 | - | 4 - 5 | 440 | 4502819 | 87 | |
| CT010305.1 | - | 4 - 5 | 433 | 87 | ||
| Y09046.1 | - | 4 - 5 | 438 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pex19
Genomic Map