Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200013045.mm.1 |
NM_028687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013045.mm.1 |
NM_028687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013045.mm.1 |
TC1200001900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013051.mm.1 |
NM_028687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013051.mm.1 |
NM_028687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013051.mm.1 |
TC1200001900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013048.mm.1 |
NM_028687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013048.mm.1 |
NM_028687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013048.mm.1 |
TC1200001900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013047.mm.1 |
NM_028687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013047.mm.1 |
NM_028687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013047.mm.1 |
TC1200001900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013050.mm.1 |
NM_028687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013050.mm.1 |
NM_028687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013050.mm.1 |
TC1200001900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013046.mm.1 |
NM_028687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013046.mm.1 |
NM_028687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013046.mm.1 |
TC1200001900.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200013049.mm.1 |
NM_028687.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013049.mm.1 |
NM_028687.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013049.mm.1 |
TC1200001900.mm.1 |
MTA TC |