Assay Details
Gene Symbol: Agxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276710.1 | NP_001263639.1 | 8 - 9 | 964 | 51 | |
NM_016702.3 | NP_057911.2 | 8 - 9 | 964 | 51 | ||
GenBank mRNA | AK149432.1 | - | 8 - 9 | 887 | 51 | |
BC025799.1 | - | 8 - 9 | 928 | 5099627 | 51 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
