Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100007965.mm.1 |
NM_001159717.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007965.mm.1 |
NM_001159717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007965.mm.1 |
NM_001159718.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007965.mm.1 |
NM_001159718.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007965.mm.1 |
NM_001159719.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007965.mm.1 |
NM_001159719.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007965.mm.1 |
NM_010891.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007965.mm.1 |
NM_010891.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007965.mm.1 |
TC0100000936.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100007966.mm.1 |
NM_001159717.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007966.mm.1 |
NM_001159717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007966.mm.1 |
NM_001159718.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007966.mm.1 |
NM_001159718.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007966.mm.1 |
NM_001159719.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007966.mm.1 |
NM_001159719.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007966.mm.1 |
NM_010891.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007966.mm.1 |
NM_010891.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100007966.mm.1 |
TC0100000936.mm.1 |
MTA TC |