Assay Details
| Gene Symbol: Ppp1r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023200.2 | NP_075689.1 | 4 - 5 | 326 | 83 | |
| GenBank mRNA | AF222867.1 | - | 4 - 5 | 326 | 83 | |
| AK009342.1 | - | 4 - 5 | 341 | 83 | ||
| AK009870.1 | - | 4 - 5 | 319 | 83 | ||
| AK031983.1 | - | 1118 | 83 | |||
| AK129024.1 | - | 4 - 5 | 343 | 83 | ||
| AK162517.1 | - | 4 - 5 | 334 | 83 | ||
| BC013524.1 | - | 4 - 5 | 327 | 4237290 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r7
Genomic Map