Assay Details
| Gene Symbol: Ndufa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024197.1 | NP_077159.1 | 6 - 7 | 768 | 72 | |
| GenBank mRNA | AK005339.1 | - | 6 - 7 | 804 | 72 | |
| AK013675.1 | - | 6 - 7 | 811 | 72 | ||
| AK046292.1 | - | 672 | 72 | |||
| AK145979.1 | - | 6 - 7 | 824 | 72 | ||
| AK183638.1 | - | 1 - 2 | 81 | 72 | ||
| BC003439.1 | - | 6 - 7 | 768 | 2647815 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufa10
Genomic Map