Assay Details
| Gene Symbol: Nr1i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC037645.1 | - | 2 - 3 | 345 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC037645.1 transcripts.
Set Membership: |
|

Nr1i3
Genomic Map