| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100012365.mm.1 |
NM_001276455.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012365.mm.1 |
NM_054087.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012365.mm.1 |
TC0100001500.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100012364.mm.1 |
NM_001276455.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012364.mm.1 |
NM_054087.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012364.mm.1 |
TC0100001500.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100012363.mm.1 |
NM_001276455.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012363.mm.1 |
NM_054087.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100012363.mm.1 |
TC0100001500.mm.1 |
MTA TC |