Assay Details
| Gene Symbol: Slc19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276455.1 | NP_001263384.1 | 3 - 4 | 1126 | 84 | |
| NM_054087.3 | NP_473428.1 | 3 - 4 | 1240 | 84 | ||
| GenBank mRNA | AF179403.1 | - | 3 - 4 | 1231 | 84 | |
| AF216204.1 | - | 3 - 4 | 1210 | 84 | ||
| AF326916.1 | - | 3 - 4 | 919 | 84 | ||
| AF360396.1 | - | 3 - 4 | 1222 | 84 | ||
| AF418986.1 | - | 3 - 4 | 1117 | 84 | ||
| AK044825.1 | - | 3 - 4 | 1241 | 84 | ||
| BC034659.1 | - | 3 - 4 | 1221 | 5359902 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc19a2
Genomic Map