Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100012593.mm.1 |
NM_001113391.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012593.mm.1 |
NM_001113391.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012593.mm.1 |
NM_001113392.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012593.mm.1 |
NM_001113392.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012593.mm.1 |
NM_001113393.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012593.mm.1 |
NM_001113393.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012593.mm.1 |
NM_031162.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012593.mm.1 |
NM_031162.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012593.mm.1 |
NR_103716.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100012593.mm.1 |
TC0100001527.mm.1 |
MTA TC |