| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100028103.mm.1 |
NM_001160208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028103.mm.1 |
NM_001160208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028103.mm.1 |
NM_001160209.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028103.mm.1 |
NM_001160209.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028103.mm.1 |
NM_025505.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028103.mm.1 |
NM_025505.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028103.mm.1 |
TC0100003421.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100028095.mm.1 |
NM_001160208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028095.mm.1 |
NM_001160208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028095.mm.1 |
NM_001160209.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028095.mm.1 |
NM_001160209.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028095.mm.1 |
NM_025505.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028095.mm.1 |
NM_025505.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028095.mm.1 |
TC0100003421.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100028096.mm.1 |
NM_001160208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028096.mm.1 |
NM_001160208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028096.mm.1 |
NM_001160209.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028096.mm.1 |
NM_001160209.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028096.mm.1 |
NM_025505.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028096.mm.1 |
NM_025505.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028096.mm.1 |
TC0100003421.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100028100.mm.1 |
NM_001160208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028100.mm.1 |
NM_001160208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028100.mm.1 |
NM_001160209.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028100.mm.1 |
NM_001160209.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028100.mm.1 |
NM_025505.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028100.mm.1 |
NM_025505.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028100.mm.1 |
TC0100003421.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100028104.mm.1 |
NM_001160209.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028104.mm.1 |
NM_001160209.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028104.mm.1 |
TC0100003421.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100028101.mm.1 |
NM_001160208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028101.mm.1 |
NM_001160208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028101.mm.1 |
NM_001160209.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028101.mm.1 |
NM_001160209.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028101.mm.1 |
NM_025505.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028101.mm.1 |
NM_025505.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028101.mm.1 |
TC0100003421.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100028105.mm.1 |
NM_001160208.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028105.mm.1 |
NM_001160208.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028105.mm.1 |
NM_001160209.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028105.mm.1 |
NM_001160209.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028105.mm.1 |
NM_025505.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028105.mm.1 |
NM_025505.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028105.mm.1 |
TC0100003421.mm.1 |
MTA TC |