Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200004299.mm.1 |
NM_001142728.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004299.mm.1 |
NM_001142728.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004299.mm.1 |
NM_001142729.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004299.mm.1 |
NM_001142729.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004299.mm.1 |
NM_030070.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004299.mm.1 |
NM_030070.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004299.mm.1 |
TC1200000622.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200004298.mm.1 |
NM_001142728.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004298.mm.1 |
NM_001142728.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004298.mm.1 |
NM_001142729.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004298.mm.1 |
NM_001142729.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004298.mm.1 |
NM_030070.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004298.mm.1 |
NM_030070.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004298.mm.1 |
TC1200000622.mm.1 |
MTA TC |