Assay Details
| Gene Symbol: Tomm40l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037170.2 | NP_001032247.1 | 4 - 5 | 552 | 90 | |
| GenBank mRNA | AK012264.1 | - | 4 - 5 | 482 | 90 | |
| AK028388.1 | - | 4 - 5 | 553 | 90 | ||
| BC058978.1 | - | 3 - 4 | 443 | 5715769 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tomm40l
Genomic Map