Assay Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153555.2 | NP_705783.1 | 12 - 13 | 1782 | 68 | |
| GenBank mRNA | AK035701.1 | - | 12 - 13 | 1681 | 68 | |
| AK098114.1 | - | 12 - 13 | 1801 | 68 | ||
| AK146825.1 | - | 12 - 13 | 1709 | 68 | ||
| BC023704.1 | - | 13 - 14 | 1901 | 5345701 | 68 | |
| BC023804.1 | - | 13 - 14 | 1836 | 5352941 | 68 | |
| BC078641.1 | - | 12 - 13 | 2694 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf8
Genomic Map