Assay Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153555.2 | NP_705783.1 | 7 - 8 | 1301 | 75 | |
| GenBank mRNA | AK035701.1 | - | 7 - 8 | 1200 | 75 | |
| AK098114.1 | - | 7 - 8 | 1320 | 75 | ||
| AK146825.1 | - | 7 - 8 | 1228 | 75 | ||
| AK162622.1 | - | 7 - 8 | 1323 | 75 | ||
| AK164799.1 | - | 7 - 8 | 1228 | 75 | ||
| AK167059.1 | - | 7 - 8 | 1177 | 75 | ||
| BC023704.1 | - | 8 - 9 | 1242 | 5345701 | 75 | |
| BC023804.1 | - | 8 - 9 | 1355 | 5352941 | 75 | |
| BC078641.1 | - | 7 - 8 | 2213 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf8
Genomic Map