Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100029271.mm.1 |
NM_013734.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029271.mm.1 |
NM_013734.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029271.mm.1 |
TC0100003565.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100029270.mm.1 |
NM_013734.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029270.mm.1 |
NM_013734.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029270.mm.1 |
TC0100003565.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100029269.mm.1 |
NM_013734.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029269.mm.1 |
NM_013734.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100029269.mm.1 |
TC0100003565.mm.1 |
MTA TC |