Assay Details
| Gene Symbol: Ilkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023343.2 | NP_075832.1 | 3 - 4 | 253 | 170 | |
| GenBank mRNA | AK005408.1 | - | 3 - 4 | 286 | 170 | |
| BC011427.1 | - | 3 - 4 | 223 | 130 | ||
| BC026953.1 | - | 3 - 4 | 253 | 5133980 | 170 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ilkap
Genomic Map