Assay Details
| Gene Symbol: Frem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172862.3 | NP_766450.2 | 17 - 18 | 8413 | 86 | |
| GenBank mRNA | AB236662.1 | - | 17 - 18 | 8413 | 86 | |
| AJ833643.1 | - | 17 - 18 | 8142 | 86 | ||
| AK031494.1 | - | 7 - 8 | 1315 | 86 | ||
| BC068185.1 | - | 7 - 8 | 1315 | 86 | ||
More Information
Set Membership: |
|

Frem2
Genomic Map