Assay Details
| Gene Symbol: Fat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183221.3 | NP_899044.3 | 8 - 9 | 7470 | 66 | |
| GenBank mRNA | DQ286572.1 | - | 8 - 9 | 7470 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fat4
Genomic Map