Assay Details
| Gene Symbol: Sox2ot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015580.2 | 2 - 3 | 257 | 74 | ||
| GenBank mRNA | BC057611.1 | - | 2 - 3 | 257 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1897 | Mir1897 |
Mirn1897, mmu-mir-1897 |
Sense | Chr. 3 - 34638464 - 34638542 on NCBI Build GRCm38 | |
| Downstream | SRY (sex determining region Y)-box 2 | Sox2 |
Sox-2, lcc, ysb |
11.4kb | Sense | Chr. 3 - 34650005 - 34652461 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Sox2ot
Genomic Map