Assay Details
| Gene Symbol: Edil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037987.3 | NP_001033076.1 | 3 - 4 | 734 | 69 | |
| GenBank mRNA | AF031524.1 | - | 3 - 4 | 842 | 69 | |
| AF031525.1 | - | 3 - 4 | 773 | 69 | ||
| AK036117.1 | - | 3 - 4 | 695 | 69 | ||
| AK046885.1 | - | 3 - 4 | 735 | 69 | ||
| AK084215.1 | - | 3 - 4 | 589 | 69 | ||
| AK135220.1 | - | 3 - 4 | 698 | 69 | ||
| AK139093.1 | - | 3 - 4 | 696 | 69 | ||
| AK141907.1 | - | 3 - 4 | 718 | 69 | ||
| AK157810.1 | - | 3 - 4 | 719 | 69 | ||
| BC056386.1 | - | 3 - 4 | 634 | 6854497 | 69 | |
More Information
Set Membership: |
|

Edil3
Genomic Map