Assay Details
| Gene Symbol: Lgr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033409.3 | NP_001028581.1 | 15 - 16 | 1539 | 82 | |
| GenBank mRNA | AK052873.1 | - | 10 - 11 | 739 | 82 | |
| AK085901.1 | - | 15 - 16 | 1498 | 82 | ||
| AK137380.1 | - | 15 - 16 | 1540 | 82 | ||
| BC141210.1 | - | 15 - 16 | 1491 | 9055822 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lgr6
Genomic Map