Assay Details
| Gene Symbol: Lgr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033409.3 | NP_001028581.1 | 14 - 15 | 1414 | 62 | |
| GenBank mRNA | AK052873.1 | - | 9 - 10 | 614 | 62 | |
| AK085901.1 | - | 14 - 15 | 1373 | 62 | ||
| AK137380.1 | - | 14 - 15 | 1415 | 62 | ||
| BC141210.1 | - | 14 - 15 | 1366 | 9055822 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lgr6
Genomic Map