Assay Details
| Gene Symbol: A930001A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040549.1 | 4 - 5 | 718 | 70 | ||
| NR_040550.1 | 3 - 4 | 503 | 70 | |||
| GenBank mRNA | AK020793.1 | - | 4 - 5 | 698 | 70 | |
| AK044583.1 | - | 3 - 4 | 504 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | carbonic anhydrase 2 | Car2 |
AI131712, CAII, Ca2, Car-2, Ltw-5, Lvtw-5 |
-77.2kb | Sense | Chr. 3 - 14886278 - 14900770 on NCBI Build GRCm38 |
| Downstream | predicted gene 9733 | Gm9733 | 292.4kb | Anti-sense | Chr. 3 - 15295103 - 15332310 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2372300 | BB280276.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2372300 | AK044583.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610712 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898671 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805412 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886434 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599214 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456796 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597142 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9852 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141275 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141275 | - | - |
| Whole Mouse Genome G4122A | A_52_P349091 | AK044583.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P349091 | AK020793.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118822 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02420607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03106653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02283929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03171142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02336525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03167296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03180909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03076716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02858753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02838317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03106894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03171144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03030741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02622781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959522 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03030753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02858745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02838320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21959615 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1431522_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431522_at | - | - |
| Mouse Genome 430 2.0 Array | 1431522_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300000441.mm.1 | NR_040549.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000441.mm.1 | NR_040549.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000441.mm.1 | NR_040550.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000441.mm.1 | NR_040550.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000441.mm.1 | TC0300000097.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431522_at | - | - |
| Mouse Expression Set 430 | 1431522_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431522_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431522_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 586069 | - | - |

A930001A20Rik
Genomic Map