| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300001703.mm.1 |
NR_029456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001703.mm.1 |
NR_029456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001703.mm.1 |
TC0300000263.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001704.mm.1 |
NR_029456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001704.mm.1 |
NR_029456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001704.mm.1 |
TC0300000263.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001698.mm.1 |
NR_029456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001698.mm.1 |
NR_029456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001698.mm.1 |
TC0300000263.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001702.mm.1 |
NR_029456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001702.mm.1 |
NR_029456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001702.mm.1 |
TC0300000263.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001701.mm.1 |
NR_029456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001701.mm.1 |
NR_029456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001701.mm.1 |
TC0300000263.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001696.mm.1 |
NR_029456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001696.mm.1 |
NR_029456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001696.mm.1 |
TC0300000263.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001697.mm.1 |
NR_029456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001697.mm.1 |
NR_029456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001697.mm.1 |
TC0300000263.mm.1 |
MTA TC |