Assay Details
| Gene Symbol: Mccc1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036093.1 | - | 517 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mccc1os| Gene Symbol: Mccc1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036093.1 | - | 517 | 76 | ||