Assay Details
| Gene Symbol: Cxcr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009911.3 | NP_034041.2 | 1 - 2 | 105 | 99 | |
| GenBank mRNA | AB000803.1 | - | 1 - 2 | 86 | 99 | |
| BB872868.1 | - | 1 - 2 | 106 | 99 | ||
| BC031665.1 | - | 1 - 2 | 92 | 3592479 | 99 | |
| BC098322.1 | - | 1 - 2 | 82 | 2864967 | 99 | |
| U59760.1 | - | 1 - 2 | 44 | 99 | ||
| X99582.1 | - | 1 - 2 | 104 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB000803.1, BC031665.1, BC098322.1, NM_009911.3, U59760.1, X99582.1 transcripts.
Set Membership: |
|

Cxcr4
Genomic Map