Assay Details
| Gene Symbol: Crb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133239.2 | NP_573502.2 | 8 - 9 | 3003 | 82 | |
| GenBank mRNA | AF406641.1 | - | 8 - 9 | 2968 | 82 | |
| AF492496.1 | - | 7 - 8 | 2844 | 82 | ||
| BC031378.1 | - | 1 - 2 | 63 | 82 | ||
| BC053331.1 | - | 3 - 4 | 1246 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crb1
Genomic Map