Assay Details
Gene Symbol: Nek7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021605.3 | NP_067618.1 | 1 - 2 | 147 | 87 | |
GenBank mRNA | AK035502.1 | - | 1 - 2 | 146 | 87 | |
AK078639.1 | - | 1 - 2 | 156 | 87 | ||
AK088173.1 | - | 1 - 2 | 196 | 87 | ||
AK149308.1 | - | 1 - 2 | 195 | 87 | ||
AK165583.1 | - | 1 - 2 | 125 | 87 | ||
BC026541.1 | - | 1 - 2 | 139 | 87 | ||
BC037697.1 | - | 1 - 2 | 124 | 3994871 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK035502.1, AK078639.1, AK088173.1, AK149308.1, AK165583.1, BC037697.1, NM_021605.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
