Assay Details
| Gene Symbol: Qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024945.1 | NP_001020116.1 | 8 - 9 | 1125 | 79 | |
| NM_023268.2 | NP_075757.1 | 8 - 9 | 1125 | 79 | ||
| GenBank mRNA | AB044284.1 | - | 8 - 9 | 1033 | 79 | |
| AK004880.1 | - | 8 - 9 | 1071 | 79 | ||
| AK083938.1 | - | 8 - 9 | 1076 | 79 | ||
| AK166839.1 | - | 8 - 9 | 1071 | 79 | ||
| AK169877.1 | - | 8 - 9 | 1078 | 79 | ||
| AK170449.1 | - | 8 - 9 | 1038 | 79 | ||
| BC034131.1 | - | 5 - 6 | 617 | 79 | ||
| BC076590.1 | - | 8 - 9 | 1079 | 30548238 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qsox1
Genomic Map