Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100026736.mm.1 |
NM_001243223.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026736.mm.1 |
NM_001243223.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026736.mm.1 |
TC0100003282.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026738.mm.1 |
NM_001243223.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026738.mm.1 |
NM_001243223.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026738.mm.1 |
TC0100003282.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026737.mm.1 |
NM_001243223.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026737.mm.1 |
NM_001243223.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026737.mm.1 |
TC0100003282.mm.1 |
MTA TC |