Assay Details
| Gene Symbol: Rgsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243223.1 | NP_001230152.1 | 3 - 4 | 191 | 93 | |
| GenBank mRNA | CJ042418.1 | - | 3 - 4 | 191 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rgsl1
Genomic Map