Assay Details
| Gene Symbol: Kcnmb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195074.1 | NP_001182003.1 | 3 - 4 | 439 | 58 | |
| GenBank mRNA | CR515784.1 | - | 1 - 2 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnmb3
Genomic Map