Assay Details
Gene Symbol: Hsd17b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010475.1 | NP_034605.1 | 3 - 4 | 470 | 60 | |
GenBank mRNA | AK143346.1 | - | 3 - 4 | 419 | 60 | |
BC125657.1 | - | 3 - 4 | 469 | 40130172 | 60 | |
BC125659.1 | - | 3 - 4 | 469 | 40130174 | 60 | |
X89627.1 | - | 3 - 4 | 470 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
